missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) acetate dimer, anhydrous, ca 46% Rh

Product Code. p-4510064 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10134970

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15956-28-2 | C8H12O8Rh2 | 441.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15956-28-2
Molecular Formula C8H12O8Rh2
Molecular Weight (g/mol) 441.99
MDL Number MFCD00003538
InChI Key VOXKIADTKKPBBP-UHFFFAOYSA-N
Synonym rhodium ii acetate, rhodium diacetate, acetic acid, rhodium 2+ salt, rhodium ii acetate dimer dihydrate, dirhodium tetraacetate, rhodium ii acetate dimer, rhodium acetate, brown, water soluble, diacetoxyrhodium
PubChem CID 122130469
SMILES C[C-]1O[Rh++]234O[C-](C)O[Rh++]2(O1)(O[C-](C)O3)O[C-](C)O4

Specifications

Melting Point 205.0°C
Color Green
Physical Form Micro-crystalline Powder
Quantity 250 mg
Merck Index 15,8311
Solubility Information Solubility in water: insoluble. Other solubilities: moderately soluble in ethanol, acetone, acetonitri, le, dmso, thf
IUPAC Name tetramethyl-2,4,6,8,9,11,12,14-octaoxa-1,5-dirhodatetracyclo[3.3.3.3¹,⁵.0¹,⁵]tetradecane-1,1,5,5-tetrakis(ylium)-3,7,10,13-tetraide
Formula Weight 441.98
Percent Purity ∼46% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium(II) acetate dimer
Show More Show Less
compliance-icons
Product Identifier
  • Rhodium(II) acetate dimer, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.