missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) acetate dimer, dihydrate, ca 43% Rh

Product Code. 10062580 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10062580

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 29998-99-0 | C8H16O10Rh2 | 478.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 29998-99-0
Molecular Formula C8H16O10Rh2
Molecular Weight (g/mol) 478.02
MDL Number MFCD00149212
Synonym Tetrakis[micron-(acetato-O:O')]diaquadirhodium

Tekniske data

Color Green
Physical Form Fine Powder
Quantity 250 mg
Linear Formula [(CH3CO2)2RhH2O]2
Fieser 08,434; 13,266; 15,278; 16,289; 17,298; 05,571
Solubility Information Solubility in water: insoluble. Other solubilities: moderately soluble in many polar solvents like eth, anol, acetone, acetic acid, acetonitrile, dmso, ni, tromethane, thf, triethyl phosphate.
Formula Weight 478.02
Percent Purity ∼43% (Rh)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Rhodium(II) acetate dimer, dihydrate, ca 43% Rh
Vis mere Vis mindre

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.