missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) 2,4-pentanedionate, Premion™, 99.99% (metals basis), Rh 25.2% min, Thermo Scientific Chemicals

445.22 € - 800.99 €

Chemical Identifiers

CAS 14284-92-5
Molecular Formula C15H21O6Rh
Molecular Weight (g/mol) 400.23
MDL Number MFCD00083144 MFCD00083144
InChI Key DGOINFUDFBWCMX-LNTINUHCSA-K
Synonym rhodium iii acetylacetonate-rh
PubChem CID 131675882
SMILES [Rh+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11300719
SDS View Documents
Thermo Scientific Alfa Aesar
010561.02
0.5 g
445.22 €
0.50g
Please to purchase this item. Need a web account? Register with us today!
11390709
SDS View Documents
Thermo Scientific Alfa Aesar
010561.03
1 g
800.99 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rhodium(III) 2,4-pentanedionate is used in catalytic reagents for organic synthesis. Rhodium(III) 2,4-pentanedionate is useful for hydrosilylation of a variety of organic substrates like alkenes, terminal or internal acetylenes, conjugated dienes, alpha, beta -unsaturated aldehydes and allylic compounds. It performs the role of a catalyst in well-known reactions such as transformation processes including hydrogenation, ethanol steam reforming, CO oxidation, and NOx reduction. Rhodium acetylacetonate, as the organic solvent-soluble source of rhodium metal ions, can be used to prepare monodisperse size/shape controlled Rh nanocrystals. Rh nanocrystal monolayers have been reported to show high activity for hydrogenation of vinylcyclohexene to selectively produce ethylcyclohexene. The size/shape controlled nanocrystals find a variety of applications in oil refining processes, chemical synthesis and fuel cell technology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14284-92-5
400.23
DGOINFUDFBWCMX-LNTINUHCSA-K
131675882
C15H21O6Rh
MFCD00083144 MFCD00083144
rhodium iii acetylacetonate-rh
[Rh+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

263°C to 264°C
Crystalline
MFCD00083144 MFCD00083144
Very soluble in chloroform; Soluble in alcohol; In soluble in water.
[Rh+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
400.23
400.23
Premion™
(metals basis)
14284-92-5
C15H21O6Rh
rhodium iii acetylacetonate-rh
DGOINFUDFBWCMX-LNTINUHCSA-K
rhodium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
131675882
99.99%
Rh 25.2% min.
Rhodium(III) 2,4-pentanedionate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'einecsNumber' : 238-192-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.