missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(III) chloride hydrate, Rh 38.0-45.5%

Product Code. p-4782099
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11341658

Brand: Thermo Scientific Alfa Aesar 011032.01

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20765-98-4 | Cl3Rh | 209.26 g/mol

Rhodium(III) chloride hydrate is used as a catalyst to prepare acetic acid. It is also used as a catalyst for reduction, oxidation of alkenes, hydration of acetylene and isomerization of alkenes. It is involved in the hydrosilylation of alfa,beta-unsaturated esters to form dimethylketene trimethylsilyl acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20765-98-4
Molecular Formula Cl3Rh
Molecular Weight (g/mol) 209.26
MDL Number MFCD00149839
InChI Key SONJTKJMTWTJCT-UHFFFAOYSA-K
Synonym Rhodium trichloride hydrate
IUPAC Name rhodium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Rh+3]

Specifications

Melting Point 100°C (decomposition)
Color Red
Physical Form Crystalline
Quantity 0.25 g
Assay Percent Range 38.5 to 45.5%
Solubility Information Soluble in alcohol and water. Slightly soluble in acetone.
Formula Weight 209.26 (Anhydrous)
Sensitivity Hygroscopic
Appearance Dark red crystals
Chemical Name or Material Rhodium(III) chloride hydrate, Rh 38.5-45.5%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.