missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(IV) oxide, Rh 75.2-77.4%, Thermo Scientific™

POA

Chemical Identifiers

CAS 12137-27-8
Molecular Formula O2Rh
Molecular Weight (g/mol) 134.904
MDL Number MFCD00016312
InChI Key KZYDBKYFEURFNC-UHFFFAOYSA-N
Synonym rhodium iv oxide, rhodium oxide rho2, rhodium dioxide, rho2
PubChem CID 82936
IUPAC Name dioxorhodium
SMILES O=[Rh]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15402827
View Documents
Thermo Scientific Alfa Aesar
047002.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15492817
View Documents
Thermo Scientific Alfa Aesar
047002.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12137-27-8
134.904
KZYDBKYFEURFNC-UHFFFAOYSA-N
82936
O=[Rh]=O
O2Rh
MFCD00016312
rhodium iv oxide, rhodium oxide rho2, rhodium dioxide, rho2
dioxorhodium

Specifications

12137-27-8
Powder
MFCD00016312
It is insoluble in aqua regia.
O=[Rh]=O
134.904
134.9
Odorless
Rhodium(IV) oxide
Black
O2Rh
rhodium iv oxide, rhodium oxide rho2, rhodium dioxide, rho2
KZYDBKYFEURFNC-UHFFFAOYSA-N
dioxorhodium
82936
Rh 75.2 to 77.4%
Hygroscopic
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-237-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.