missing translation for 'onlineSavingsMsg'
Learn More

Rhodizonic acid, disodium salt, 98%, Thermo Scientific Chemicals

Product Code. 15119183
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 523-21-7
Molecular Formula C6Na2O6
Molecular Weight (g/mol) 214.04
MDL Number MFCD00001650
InChI Key SXWPCWBFJXDMAI-UHFFFAOYSA-L
Synonym sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate
PubChem CID 68225
SMILES [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O

Specifications

Melting Point >300.0°C
Color Gray-Green to Green
Physical Form Fine Crystalline Powder
Quantity 100 g
Assay Percent Range 98%
Beilstein 08,536
Merck Index 15,8799
Solubility Information (0.2% in water) Clear
IUPAC Name disodium;3,4,5,6-tetraoxocyclohexene-1,2-diolate
Formula Weight 214.04
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Appearance (0.2% aq. soln.) clear
Chemical Name or Material Rhodizonic acid, disodium salt
Show More Show Less
Product Identifier
  • Rhodizonic acid, disodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.