missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIAM Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547911
This item is not returnable.
View return policy
Description
This antibody detects recombinant human RIAM/APBB1IP in direct ELISAs. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
RIAM Polyclonal antibody specifically detects RIAM in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
RIAM | |
Polyclonal | |
Unconjugated | |
APBB1IP | |
9930118P07Rik; amyloid beta (A4) precursor protein-binding, family B, member 1 interacting protein; amyloid beta A4 precursor protein-binding family B member 1-interacting protein; amyloid beta precursor protein binding family B member 1 interacting protein; APBB1-interacting protein 1; Apbb1ip; INAG1; Prel1; PREL-1; proline rich EVH1 ligand 1; proline-rich EVH1 ligand 1; Proline-rich protein 48; proline-rich protein 73; Prp48; Rap1-GTP-interacting adapter molecule; Rap1-GTP-interacting adaptor molecule; Rap1-interacting adaptor molecule; RARP1; RARP-1; retinoic acid-responsive proline-rich protein 1; RIAM; RIAMRARP-1; zgc:63968 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
54518, 54519 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q7Z5R6, Q8R5A3 | |
APBB1IP | |
E. coli-derived recombinant human RIAM/APBB1IP Gly2-Ser89 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction