missing translation for 'onlineSavingsMsg'
Learn More
Ribavirin, 98%, Thermo Scientific Chemicals Product Code.: 15599513

Ribavirin, 98%, Thermo Scientific Chemicals

Product Code. 15599513
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15599513

Brand: Thermo Scientific Acros 460480010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ribavirin, CAS# 36791-04-5, is an antiviral agent and a guanosine analog, which stops viral RNA synthesis and mRNA capping.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ribavirin is a broad-spectrum antiviral nucleoside analog
  • It inhibits viral genome synthesis

Applications

  • Ribavirin is an antiviral compound
  • Its metabolite, ribavirin 5'-phosphate, inhibits inosine monophosphate dehydrogenase (IMP), which is involved in GTP biosynthesis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36791-04-5
Molecular Formula C8H12N4O5
Molecular Weight (g/mol) 244.2
InChI Key IWUCXVSUMQZMFG-AFCXAGJDSA-N
Synonym ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere
PubChem CID 37542
ChEBI CHEBI:63580
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide
SMILES C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N

Specifications

Ignition Residue 0.25% max.
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: 142 mg/mL @25°C.
Formula Weight 244.2
Percent Purity 98%
Chemical Name or Material Ribavirin
compliance-icons
Product Identifier
  • Ribavirin
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ribavirin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.