missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Thermal Transfer Printer Ribbon
With high chemical resisntance at high temperatures
Brand: Brady™ 804777
Code : 50
Additional Details : Weight : 0.04680kg
Description
- Excellent solvent resistance
- Ribbon ink side out
Specifications
12.7 mm I.D. | |
70 m x 65 mm | |
Black 6400 Series Thermal Transfer Printer Ribbon | |
Ink side out |