missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Riboflavin, 98%
38.40 € - 400.00 €
Chemical Identifiers
CAS | 83-88-5 |
---|---|
Molecular Formula | C17H20N4O6 |
Molecular Weight (g/mol) | 376.37 |
MDL Number | MFCD00005022 |
InChI Key | AUNGANRZJHBGPY-QTZZOOGMNA-N |
Synonym | riboflavin, vitamin b2, lactoflavin, riboflavine, vitamin g, lactoflavine, --riboflavin, beflavin, beflavine, flavaxin |
PubChem CID | 71310809 |
SMILES | CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10593871
|
Thermo Scientific Chemicals
132350250 |
25 g | Glass bottle |
38.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10438870
|
Thermo Scientific Chemicals
132351000 |
100 g | Glass bottle |
105.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10135783
|
Thermo Scientific Chemicals
132355000 |
500 g | Glass bottle |
400.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
83-88-5 | |
376.37 | |
AUNGANRZJHBGPY-QTZZOOGMNA-N | |
71310809 |
C17H20N4O6 | |
MFCD00005022 | |
riboflavin, vitamin b2, lactoflavin, riboflavine, vitamin g, lactoflavine, --riboflavin, beflavin, beflavine, flavaxin | |
CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1 |
Specifications
83-88-5 | |
290.0°C | |
1.5% max. (105°C) | |
98% | |
MFCD00005022 | |
− 125.00 (20.00°C c=0.5,0.05 M NaOH) | |
riboflavin, vitamin b2, lactoflavin, riboflavine, vitamin g, lactoflavine, --riboflavin, beflavin, beflavine, flavaxin | |
97.5% min. (lambda max. 444nm) (A (1%/1 cm) 328,on dried substance) | |
CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1 | |
376.37 | |
376.36 | |
Powder |
0.31 to 0.33 (A373/A267), 0.36 to 0.39 (A444/A267) | |
Yellow to Orange | |
Authentic | |
C17H20N4O6 | |
15, 8325 | |
-125 | |
Solubility in water: 0.07g/L (20°C). Other solubilities: very soluble in diluted alkalies,,slightly soluble in ethanol and nacl solutions,,insoluble in ethers,chloroform,acetone,benzene,and lipid solvents | |
AUNGANRZJHBGPY-QTZZOOGMNA-N | |
7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione | |
71310809 | |
98% | |
Riboflavin |
Safety and Handling
missing translation for 'einecsNumber' : 201-507-1
RUO – Research Use Only