missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RiboLock RNase Inhibitor (40 U/μL)

RiboLock RNase Inhibitor specifically inhibits the activity of RNases A, B, and C, protecting RNA under a variety of reaction conditions.

Brand:  Thermo Scientific™ EO0382

 View more versions of this product

Product Code. 11581505

  • 289.00 € / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

RiboLock RNase Inhibitor inhibits the activity of RNases A , B, and C by binding them in a noncompetitive mode at a 1:1 ratio. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H.

  • Does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 as well as prokaryotic RNases I and H
  • Performs under a wide range of reaction conditions
  • Protects RNA from degradation at temperatures up to 55°C
Quality Control:
  • The absence of ribonucleases, endo-, exodeoxyribonucleases and phosphatases and ribonucleases confirmed by appropriate quality tests
  • Functionally tested in RNA and cDNA synthesis
Source:
  • E.coli cells with a cloned gene encoding mammalian ribonuclease inhibitor
Molecular Weight:
  • 49.6kDa monomer Definition of Activity Unit:
    • One unit of the RiboLock RNase Inhibitor inhibits the activity of 5ng RNase A by 50%
    • Inhibitor activity is assayed in the following mixture: 100mM Tris-HCl (pH 7.5), 1.2mM EDTA, 0.1mg/mL BSA, 100ng/mL RNase A, 0.1mg/mL [3H]-RNA, 50mg/mL yeast RNA, 8mM DTT
    Storage Buffer:
    • The enzyme is supplied in: 20mM HEPES-NaOH (pH 7.5), 50mM NaCl, 8mM DTT, 0.5mM ELUGENT Detergent and 50% (v/v) glycerol
    Inhibition and Inactivation
    • Inhibitors: common denaturants (SDS, urea and all oxidizing reagents (p-chloromercuribenzoate, dissolved oxygen, ions in their higher oxidation states) strongly inhibit RiboLock RNase Inhibitor and release the RNase bound
    • Inactivated by heating at 75°C for 10 min. Residual activity detectable after 10 min. heating at 70°

    Recommended for:

    • Inhibition of RNA degradation in the following: in vitrotranscription (1), cDNA synthesis (2), in vitrotranslation (3), isolation of mammalian cell fractions that contain mRNA-protein complex
    • RNA purification and storage
    • Separation and identification of specific ribonuclease activities
    • Studies of tumor suppression

    Note:

    • DTT provided in the Storage Buffer ensures stability during long term storage, but is not necessary for inhibitor activity
    • Recommended concentration 1u/μl of a reaction mixture
    TRUSTED_SUSTAINABILITY
Specifications

Specifications

40 U/μL
4 x 2,500 units
RiboLock™
Storage Buffer
RNase Inhibitor
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.