Learn More
Novus Biologicals™ Ribosomal protein L26 Overexpression Lysate
Brand:  Novus Biologicals™ NBL1-15517
Product Code. 18119479
  • 700.00 € / 0.10mg
Explore more special offers
Description
Description
The Ribosomal protein L26 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC29922 and based on accession number NM_987. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens ribosomal protein L26 (RPL26), mRNA.
Native