Learn More
Epredia™ Parapel™ Paraffin Repellent
Protect against excess buildup of wax on countertops and instrumentation with Epredia™ Parapel™ Paraffin Repellent.
Brand: Epredia™ 2300
This item is not returnable.
View return policy
Description
- Low odor
- Reduces daily cleaning times
- Will not leave behind oily residue
- Will not remove finish from surfaces
Specifications
4 oz. pump bottle; 6/CS |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- ParaPel
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P331-Do NOT induce vomiting.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P273-Avoid release to the environment.
- P370+P378-In case of fire: Use to extinguish.
- P391-Collect spillage.