Learn More
Epredia™ Cytoseal™ Mountant
Prevent cracking, fading and discoloration of stained specimens with Cytoseal™ mounting media.
Brand: Epredia™ 8310-4
78.00 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
Description
- Cytoseal 60 mountant is a low-viscosity medium that dries quickly and allows for rapid, even spreading which virtually eliminates air bubbles
Ordering Information:
Packaged in an easy-to-use dispenser with a twist-open/twist-closed cap.
Specifications
Cytoseal 60 | |
Mountant | |
4 oz. |
Safety and Handling
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : toluene, acrylic resin, butyl benzyl phthalate, 2,6-ditertbutyl-p-cresol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.