Learn More
Epredia™ HistoGel™ Specimen Processing Gel
Encapsulate and retain even small, friable and viscous specimens during histological processing with HistoGel™ Specimen Processing Gel.
Brand:  Epredia™ HG-4000-144
1042.80 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 12608606
  • 1215.00 € / Pack of 144
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Penetrates specimen without allowing specimen to escape from gel
  • Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
  • Eliminates unwanted discoloration around specimens on slides
  • Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
Specifications
Specifications
Vial
Specimen Processing Gel
144 Pk.