Learn More
Epredia™ Richard-Allan Scientific™ Iron Stain, Nuclear Red Thiazin stain solution
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Richard-Allan Scientific™ Iron Stain.
Brand:  Epredia™ 88015
 View more versions of this product
Product Code. 12583637
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Useful in study of iron deficiency anemia, hemochromatosis and hemosiderosis
  • Iron stains blue, nuclei stains pink-red, and cytoplasm stains pale pink
Specifications
Specifications
Nuclear Fast Red