missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Mounting Medium

Store slides for prolonged periods of time without fading of stains with Epredia™ Mounting Medium.

64.85 € - 118.00 €

Specifications

Product Type Mounting Medium
View More Specs

Products 2
Product Code Brand Volume (English) Quantity Price Quantity & Availability  
Product Code Brand Volume (English) Quantity Price Quantity & Availability  
12657746
View Documents
Epredia™
4111
1 pt. 2/Cs.
118.00 €
940mL
Please to purchase this item. Need a web account? Register with us today!
12667746
View Documents
Epredia™
4112
4 oz. 6/Cs.
64.85 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Rapid drying
  • Toluene-based
  • Optically clear
Ordering Alerts: Packaged in two configurations for convenience: a 1-pt. regular bottle or a 4-oz. plastic squeeze bottle with a twist-open/twist-close cap.
Specifications

Specifications

Mounting Medium
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Germ cell mutagenicity Category 2

Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : toluene, acrylic resin

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.