Learn More
Epredia™ Neg-50™ Frozen Section Medium
Yield high-quality sections down to -50°C without chatter using water-soluble Epredia™ Neg-50™ Frozen Section Medium. Comes in clear and five additional colors with all leaving no residue after staining.
Brand:  Epredia™ 6502O
45.91 EUR valid until 2024-11-29
Use promo code "21706" to get your promotional price.
 View more versions of this product
Product Code. 12698086
  • 60.50 € / Pack of 2
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Yield high-quality sections down to -50°C without chatter using Epredia; Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
  • Buffered to ensure proper alkalinity (pH = 8.0); leaves no residue after staining
  • Specially formulated to prevent tissue separation from the slide
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.
Specifications
Specifications
Orange
2/Cs.
4 oz. per bottle