missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Resolve™ Immersion Oil
Eliminate toxicity and disposal problems with the Epredia™ Resolve™ Immersion Oil.
71.80 € - 256.00 €
Product Code | Brand | Quantity | Viscosity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Viscosity | Price | Quantity & Availability | ||||
12540037
|
Epredia™
M2000 |
0.5 oz. | Low |
88.75 €
Pack of 12 |
Estimated Shipment: 16-01-2025
Log in to see stock. |
||||
12590037
|
Epredia™
M4004 |
4 oz. | High |
256.00 €
Pack of 12 |
Estimated Shipment: 31-12-2024
Log in to see stock. |
||||
12550037
|
Epredia™
M2004 |
0.5 oz. | High | N/A | |||||
Description
- Manufactured using a proprietary process that removes interfering substances and will not damage valuable optics or irreplaceable slide specimens
- Packaged conveniently in four sizes
Product Details:
- Available in both low and high viscosity
- Non-drying and non-hardening
- Refractive index of 1.5150
Specifications
1.515 |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H335-May cause respiratory irritation.
- H371-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : white mineral oil, terphenyls, polyphenyls, polybutane, hydrogenated terphenyls