Learn More
Epredia™ Richard-Allan Scientific™ Three-Step Stain Kit
Obtain results on par with Wright-Giemsa with Epredia™ Richard-Allan Scientific™ Three Step Stain.
Brand:  Epredia™ 3313
 View more versions of this product
Product Code. 10425339
  • 275.00 € / Each
Explore more special offers
Includes: A package insert is included with easy-to-follow instructions.
Description
Description
  • Stain can be used to detect H. Pylori.
  • Set contains easy-to-follow instructions
  • Product is available in one-gallon containers to facilitate use on automated strainers
  • Set contains three 500mL bottles consisting of Fixative Solution, Solution A(Eosin-Y), and Solution B(Azure A and Methylene Blue)
Order Info
Components are also available in one-gallon containers to facilitate use on automated stainers.
Specifications
Specifications
1 gal.
Solution A
Stain set provides results equal to wright-giemsa
A package insert is included with easy-to-follow instructions