missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Richard-Allan Scientific™ Three-Step Stain Kit
Obtain results on par with Wright-Giemsa with Epredia™ Richard-Allan Scientific™ Three Step Stain.
Brand: Epredia™ 3300
92.40 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
- Stain can be used to detect H. Pylori.
- Set contains easy-to-follow instructions
- Product is available in one-gallon containers to facilitate use on automated strainers
- Set contains three 500mL bottles consisting of Fixative Solution, Solution A(Eosin-Y), and Solution B(Azure A and Methylene Blue)
Specifications
Epredia Three Step Stain Kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- three step fixative
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P235-Keep cool.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P363-Wash contaminated clothing before reuse.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methyl alcohol, C.I.basic green 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction