missing translation for 'onlineSavingsMsg'
Learn More
Riluzole, 98%, Thermo Scientific Chemicals Product Code.: 15548241

Riluzole, 98%, Thermo Scientific Chemicals

Product Code. 15548241
1 g
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy

Product Code. 15548241

Brand: Thermo Scientific Acros 459360010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1744-22-5
Molecular Formula C8H5F3N2OS
Molecular Weight (g/mol) 234.2
InChI Key FTALBRSUTCGOEG-UHFFFAOYSA-N
Synonym riluzole, rilutek, 2-amino-6-trifluoromethoxy benzothiazole, 6-trifluoromethoxy benzo d thiazol-2-amine, 6-trifluoromethoxy-1,3-benzothiazol-2-amine, riluzolum, riluzol, 2-amino-6-trifluoromethoxybenzothiazole, riluzole hcl, riluzol inn-spanishShow MoreShow Less
PubChem CID 5070
ChEBI CHEBI:8863
IUPAC Name 6-(trifluoromethoxy)-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1OC(F)(F)F)SC(=N2)N

Specifications

Melting Point 114.0°C to 118.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ether,acetone,dmf,chlorofo,rm
Formula Weight 234.2
Percent Purity 98%
Chemical Name or Material Riluzole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464627 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number459360100,459360010,459360050
Lot NumberA0424593 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number459360100,459360010,459360050
Lot NumberA0397065 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number459360100,459360010,459360050
compliance-icons
Product Identifier
  • Riluzole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Riluzole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.