Learn More
RIMS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15088-1-AP-20UL
 View more versions of this product
Product Code. 16884094
  • 145.00 € / 20µL
Explore more special offers
Description
Description
Rab3-interacting molecules (RIMs) are synaptic proteins are synaptic proteins necessary for neural transmission and plasticity. While both Rim1 and Rim 2 are thought to be effector proteins for Rab3, binding to Rab3 on synaptic vesicles in a GTP-dependent manner, less is known of Rim3. Expression of Rim3 in PC12 cells induced a significant increase in calcium-triggered exocytosis, with no appreciable change in the baseline release, suggesting that it plays a role in the regulation of exocytosis. Rim3 protein localizes primarily to neuronal dendrites and the postsynaptic densities, as opposed to Rim1 which is found in presynapse locations, indicating that Rim3 may contribute to synapse transmission and plasticity.
Specifications
Specifications
RIMS3
Polyclonal
Unconjugated
RIMS3
KIAA0237, NIM3, Rab 3 interacting molecule 3, RIM 3, RIM3, RIM3 gamma, RIMS3
Rabbit
Antigen Affinity Chromatography
RUO
242662, 65025, 9783
-20°C
Liquid
Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80U57, Q9JIR3, Q9UJD0
Rims3
RIMS3 Fusion Protein Ag7171
20 μL
Primary
Mouse, Rat, Human