missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIMS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | RIMS4 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16839574
|
Proteintech
20091-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16829574
|
Proteintech
20091-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to RIMS4.
Regulates synaptic membrane exocytosis.Specifications
RIMS4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P60191, Q8CIX1, Q9H426 | |
140730, 241770, 266976 | |
Peptide | |
Primary | |
-20°C | |
RIMS4 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C20orf190, Rab3 interacting molecule 4, RIM 4, RIM4, RIM4 gamma, RIMS4 | |
RIMS4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title