missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Ring With Bossehead

Manufactured from zinc alloy steel.

Brand:  Fisherbrand™ 2013/5

 View more versions of this product

Product Code. 10022054

  • 27.85 € / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Clamping nuts for rods up to 16mm in diameter
  • Accessory item
  • Stem length to the center of the bosshead is approximately 60mm
  • Bosshead in alloy, diecast on to the ring stem and fitted with a blue plastic headed nickel plated steel screw

Specifications

Specifications

75 mm
With Bossehead
Ring With Bossehead
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.