missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Ringer's Solution, sodium free, lactate-buffered
Specifications
Chemical Name or Material | Ringer's Solution |
---|---|
Physical Form | Liquid |
Synonym | dihydrogen oxide, dihydrogen monoxide |
Packaging | Plastic bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ringer's Solution | |
Liquid | |
dihydrogen oxide, dihydrogen monoxide | |
Plastic bottle |
Sicherheit und Handhabung
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only