missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Ringers 085 R-Flex Impact Gloves
Click to view available options
Size:
10
11
12
3X-Large
7
8
9
Unit Size:
1 pair
Description
- The Ringers 085 R-Flex Impact glove with Nitrile Half Dip and Fleece Lining combines protection against impact hazards and the highest levels of cut resistance, with added warmth for application in cold environments.
- A breathable knit shell developed with Ringers' F3 Technology™ and patented TPR design for superior dexterity and comfort.
- The sandy finish palm ensures secure grip on wet and dry surfaces, and touchscreen compatible index, middle and thumb tips allow technology interaction.
Specifications
Specifications
| Applications | Industrial |
| Coating Material | Nitrile |
| Color | Multicolored |
| Cut Resistance Rating | ANSI A6 |
| Hand | Specific |
| Latex Free | Yes |
| Material | HPPE, Nitrile |
| Product Type | Mechanical Protection Gloves |
| Size | 9 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction