missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rinsing Solution I, 55 to 57 wt. % water basis, Honeywell Riedel-de Haën™
CHROMASOLV™, LabReady™ Solvent Blend, rinsing agent for LC-MS
Brand: Honeywell-Riedel-de Haen 34689-1L
This item is not returnable.
View return policy
Description
- High purity chromatography solvents
- Suitability tested for specified use from HPLC up to UHPLC-MS applications.
Specifications
Rinsing Solution I | |
22°C (71.6°F) | |
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol |
1 L | |
Glass Bottle | |
55 to 57 wt. % water basis (Karl Fischer) |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Rinsing Solution I
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Propan-2-ol