missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | RIPK2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16834524
|
Proteintech
15366-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16824524
|
Proteintech
15366-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serine/threonine/tyrosine kinase that plays an essential role in modulation of innate and adaptive immune responses. Upon stimulation by bacterial peptidoglycans, NOD1 and NOD2 are activated, oligomerize and recruit RIPK2 through CARD-CARD domains. Contributes to the tyrosine phosphorylation of the guanine exchange factor ARHGEF2 through Src tyrosine kinase leading to NF-kappaB activation by NOD2. Once recruited, RIPK2 autophosphorylates and undergoes ′Lys-63′-linked polyubiquitination by E3 ubiquitin ligases XIAP, BIRC2 and BIRC3. The polyubiquitinated protein mediates the recruitment of MAP3K7/TAK1 to IKBKG/NEMO and induces ′Lys-63′-linked polyubiquitination of IKBKG/NEMO and subsequent activation of IKBKB/IKKB. In turn, NF-kappa-B is released from NF-kappa-B inhibitors and translocates into the nucleus where it activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Plays also a role during engagement of the T-cell receptor (TCR) in promoting BCL10 phosphorylation and subsequent NF-kappa-B activation.Specifications
RIPK2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O43353, P58801 | |
192656, 362491, 8767 | |
RIPK2 Fusion Protein Ag0909 | |
Primary | |
-20°C | |
RIPK2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CARD3, CARDIAK, CCK, GIG30, Receptor interacting protein 2, RICK, RIP 2, RIP2, RIPK2 | |
Ripk2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title