missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK3/RIP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 52 publications
266.00 € - 688.00 €
Specifications
| Antigen | RIPK3/RIP3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.1-0.5 ug/ml, ELISA 1:100 - 1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunoprecipitation 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml, Gel Supershift Assay, Knockout Validated, KnockDown Validated |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gel Shift, Immunoassay, KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18788803
|
Novus Biologicals
NBP1-77299SS |
0.025 mg |
266.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18731624
|
Novus Biologicals
NBP1-77299 |
688.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
RIPK3/RIP3 Polyclonal specifically detects RIPK3/RIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Supershift Assay, Knockout Validated, Knockdown Validated.Specifications
| RIPK3/RIP3 | |
| Western Blot 0.1-0.5 ug/ml, ELISA 1:100 - 1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunoprecipitation 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml, Gel Supershift Assay, Knockout Validated, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 | |
| RIPK3 | |
| IgG | |
| Affinity Purified | |
| Mouse RIPK3/RIP3 Antibody has one isoform (486aa, 53 kDa). Human RIP3 has 3 isoforms, including isoform 1 (518aa, 57 kDa), isoform 2 (252aa, 28 kDa) and isoform 3 (231aa, 25 kDa). Rat RIP3 also has one isoform (478aa, 52 kDa). NBP1-77299 can detect can detect isoforms of mouse and rat as well as human isoform 1. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Gel Shift, Immunoassay, KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| AAF03133 | |
| 11035 | |
| RIPK3/RIP3 Antibody was made to a 14 amino acid peptide near the carboxy terminus of murine RIP3. The immunogen is located within the last 50 amino acids of RIP3. Amino Acid Squence: AQFGRGRGWQPFHK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 53 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title