Learn More
Ritter Plastic™ Riplate™ SW 2 mL Capmat
Developed for optimal use of capacity when it comes to sample storage with 96 wells.
Brand:  Ritter Plastic™ 43001-0022
Product Code. 16650362
  • 295.00 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Thin-walled and parallel cavities ensure a maximum of contamination-free sample storage per well
  • With alphanumeric marking
  • Plates can be closed by hot-sealing
  • Manufactured out of pure polypropylene without lubricants for an immaculate and secure analysis
  • Relatively high volume allows closing of riplate with capmats without any liquid displacement and cross-contamination towards comparable plates with lower volume
Specifications
Specifications
Polypropylene
20/Pk.
Capmat
96
Closing square wells
Riplate SW 96
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ritter Plastic™ Riplate™ SW 2 mL Capmat > SW 96, 2 mL