missing translation for 'onlineSavingsMsg'
Learn More

RJR 2403 oxalate, Tocris Bioscience™

Product Code. 30001181
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

α4β2 selective nicotinic agonist

A neuronal nicotinic receptor agonist, showing high selectivity for the α4β2 subtype (Ki values are 26 and 36000 nM for α4β2 and α7 receptors respectively). Active in vivo.

Chemical Identifiers

CAS 220662-95-3
Molecular Formula C12H16N2O4
Molecular Weight (g/mol) 252.27
InChI Key WTIZFOAIQXMQHC-DPZBITMOSA-N
Synonym rjr-2403 oxalate, oxalic acid; rivanicline, e-n-methyl-4-3-pyridinyl-3-buten-1-amine oxalate, e-n-methyl-4-3-pyridinyl-3-bute n-1-amine oxalate, e-n-methyl-4-pyridin-3-yl but-3-en-1-amine oxalate, e-metanicotine oxalate;rivanicline oxalate;rjr 2403 oxalate, oxalic acid-3e-n-methyl-4-pyridin-3-yl but-3-en-1-amine 1/1
PubChem CID 23298272
IUPAC Name (E)-N-methyl-4-pyridin-3-ylbut-3-en-1-amine;oxalic acid
SMILES CNCCC=CC1=CN=CC=C1.C(=O)(C(=O)O)O

Specifications

Quantity 10 mg
Formula Weight 252.27
Percent Purity >99%
Chemical Name or Material RJR 2403 oxalate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.