missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNase AWAY™ Surface Decontaminant
GREENER_CHOICE

Eliminate RNase and DNA from laboratory surfaces. This product reduces dependency on carcinogenic DEPC treatments and saves time spent baking glassware.

47.80 € - 187.00 €

Specifications

pH 11
Composition Sodium Hydroxide with Water
View More Specs

Products 3
Product Code Brand Container Type Volume (English) Volume (Metric) Price Quantity & Availability  
Product Code Brand Container Type Volume (English) Volume (Metric) Price Quantity & Availability  
11580095
GREENER_CHOICE
View Documents
Thermo Scientific™
7000
Bottle 8.5 oz. 250 mL
47.80 €
250mL
In Stock
to see stock.
Edge
Add to basket
 
10666421
GREENER_CHOICE
View Documents
Thermo Scientific™
7002
Spray Bottle 16 oz. 473 mL
65.90 €
475mL
In Stock
to see stock.
Edge
Add to basket
 
11952385
GREENER_CHOICE
View Documents
Thermo Scientific™
7005-11
Bottle 1.06 gal. 4 L
187.00 €
4 L
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Eliminate RNase and DNA from laboratory surfaces. Thermo Scientific™ RNase AWAY™ is ideal for decontaminating apparatus, bench-tops, glassware and plasticware. It reduces dependency on carcinogenic DEPC treatments and saves time needed to bake glassware.

  • Use on pipettes, gel boxes, or RNA or DNA prep areas
  • Leaves no residue to interfere with gel polymerization or staining
  • Chemically stable and nonabrasive
  • Contains no strong acids
Specifications

Specifications

11
Sodium Hydroxide with Water
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.