missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ RNase Erase™

Spray on and rinse off for an Rnase-free surface. RNase Erase™ is precisely formulated to completely remove Rnase contamination from bench tops, glass, stainless steel and plastic.

Brand:  MP Biomedicals™ 112440204

Product Code. 11363179

  • 141.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Clear, colorless solution
  • Precisely formulated
  • Ribonuclease decontaminant
  • Non-toxic
  • Packaged with adjustable spray bottle for ease of use
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Room Temperature (15°C to 30°C)
Solution
500 mL
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.