Learn More
Rnase-free Water, non DEPCAbnova™
Product Code. 16032048
Click to view available options
Quantity:
5 x 1 mL
2 x 50 mL
500 mL
RNase-free Water is suitable to experiment and research of RNA.
  • The RNase-free Water, non DEPC was prepared from highly purified water by ultrafiltration without DEPC treatment.
  • The product was confirmed to be free from RNase and DNase.
  • Apperance is clear liquid.
Specifications
Quantity 5 x 1 mL