missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF34 Polyclonal Antibody

Brand: Thermo Scientific PA5113677
This item is not returnable.
View return policy
Description
Specifications
| RNF34 | |
| Polyclonal | |
| Unconjugated | |
| RNF34 | |
| AW061037; AW536122; BC004042; C88279; CARP1; CARP-1; Caspase regulator CARP1; caspases-8 and -10-associated RING finger protein 1; E3 ubiquitin-protein ligase RNF34; FLJ21786; FYVE-RING finger protein MOMO; hRFI; Human RING finger homologous to inhibitor of apoptosis protein; Momo; phafin 1; phafin-1; RFI; RIF; RIFF; ring finger protein 34; ring finger protein 34, E3 ubiquitin protein ligase; RING finger protein MOMO; RING finger protein RIFF; RING-type E3 ubiquitin transferase RNF34; RNF34 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80196 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q969K3 | |
| RNF34 | |
| Synthetic peptide directed towards the middle region of human RNF34. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction