Learn More
STEMCELL Technologies RoboSep™ Human NK Cell Isolation Kit
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
Brand:  STEMCELL Technologies 17955RF
Encompass
Product Code. 17177821
  • 1285.50 € / 1 set
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Human NK Cell Isolation Kit is designed to isolate NK cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
  • This product can be used in place of the EasySep™ Human NK Cell Enrichment Kit (Catalog #19055) for even faster cell isolations
Specifications
Specifications
NK Cells
Cell Isolation
1 x 109 cells