Learn More
SP Bel-Art™ Robotic Sterile Pipette Tips
Designed for use with automated liquid handling equipment
Brand:  SP Bel-Art™ F37860-2100
 View more versions of this product
Product Code. 17214984
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Sterile, disposable, filtered pipette tips can be used in robotic systems
    • Filtered to prevent contamination of pipette arm
  • Conductive tip models detect liquid level when pipetting conductive liquids
    • Ensures repeatable volume accuracy
  • DNase, RNase, pyrogen free, manufactured in Class 8 clean room
  • CE marked
Specifications
Specifications
1000 μL
Sterile
CE marked
16 racks x 96 tips
Robotic pipette tips
Single-use
Polypropylene
Filtered
Clear
For automated liquid handling equipment
1536
No
Tecan Genesis Freedom, Freedom EVO and Miniprep with LiHa