missing translation for 'onlineSavingsMsg'
Learn More

Testo™ Type K Air Probe

Fast, robust and easy to use for determination of the air temperature in the environment being measured.

Brand:  Testo™ 0602 1793

Product Code. 10453873

  • 118915.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Thermocouple type K
  • Short response time thanks to the exposed sensor
  • Can be connected to a wide range of measuring instruments
  • Measuring range of −60° to +400°C
  • Measurement result can be optimized by moving the probe through the air at approximately 1.5m/s
  • Includes fixed 1.2m cable
Specifications

Specifications

4 mm
115 mm
400°C
-60°C to +400°C
1.2 m
200 sec. reaction time
-60°C
Class 2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers