missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ROI & Background Calibration Kit, Fast 96-well

Product Code. 15870403 Shop All Applied Biosystems Products
Click to view available options
Quantity:
2 x 96 well plates
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 15870403

Brand: Applied Biosystems™ 4432638

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to maintain your real-time PCR instrument with Fast 96-well block.

Region of interest (ROI) and background calibration should be run after lamp change.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C ± 5°C.

Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Reaction Speed Fast
Product Type Background Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 2 x 96 well plates
Calibration Type Spectral Calibration
PCR Method qPCR
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.