missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rolitetracycline, MedChemExpress
MedChemExpress Rolitetracycline, a derivative of tetracycline, is a broad-spectrum antibiotic. Rolitetracyclin has a role as a protein synthesis inhibitor, an antiprotozoal drug and a prodrug.
Brand: Medchem Express HY-18257
This item is not returnable.
View return policy
Description
Rolitetracycline, a derivative of tetracycline, is a broad-spectrum antibiotic. Rolitetracyclin has a role as a protein synthesis inhibitor, an antiprotozoal drug and a prodrug.Specifications
Rolitetracycline | |
Light Brown | |
C27H33N3O8 | |
H2O : 10 mg/mL (18.95 mM; Need ultrasonic and warming) ∣Ethanol : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble) | |
527.57 | |
95.0% | |
Solid |
751-97-3 | |
Research | |
10 mg | |
O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])C[C@]3([H])[C@](C)(O)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4)NCN5CCCC5 | |
527.57 | |
Research | |
COVID-19-immunoregulation |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only
Spot an opportunity for improvement?Share a Content Correction