Learn More
Brady™ Roll Form Linerless Pipe Markers, with Pictograms - Gas
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand:  Brady™ N007510
Product Code. 15379085
  • 330.00 € / 1 roll
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, this saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
  • The design and layout of Brady's roll form markers ensure maximum visibility from all sides.
  • Parts supplied with a Black arrow printed on white background.
Specifications
Specifications
FREON 22 100X33RL-T3-LL-P15
Permanent Acrylic
Black on Ochre
Roll
Pipe Marker
Pipe and valve marking
220 Markers/Roll
-40°F to +284°F
0.0047 in.
33 m x 127
108.27 ft.
5 in.
5.91 in.
13 mm
Roll form linerless Pipe Markers, with pictograms - Gas - Freon 22
Silicone
Warning
Polyester
Freon 22
English
220 Pk.
-40°C to +140°C
0.119 mm
33 m x 127 mm x 150 mm
33 m
127 mm
150 mm