Learn More
Thermo Scientific™ Rollout Drawers
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire refrigerator shelving.
Brand:  Thermo Scientific™ 7446
 View more versions of this product
Product Code. 13203219
  • 4271.25 € / 1 unit
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.
  • Stainless-steel drawers
  • Adjustable with full extension
Ordering Information:
  • Factory installed; must be specified when ordering the refrigerator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complete SS Drawer Set
326 L
7
11.5 cu. ft.
Stainless Steel (Drawer)
11.5 cu. ft. (326L) Freezers