Learn More
STEMCELL Technologies RosetteSep™ DM-L Density Medium
Density gradient medium for use with RosetteSep™ CD3+ T cell, CD4+ T cell, CD8+ T cell, and B cell enrichment cocktails - 100 mL
Brand:  STEMCELL Technologies 15705
Encompass_Preferred
Product Code. 17127711
  • 106.65 € / 100mL
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • RosetteSep™ DM-L is a density gradient medium designed specifically for use with RosetteSep™ cocktails for the enrichment of specific human lymphocyte populations (CD3+ T cells, CD4+ T cells, CD8+ T cells, and B cells) from whole blood. 
  • RosetteSep™ DM-L has a density of 1.081 g/mL. 
Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled “For Research Use Only”. Please note there have been no changes to the form, function and quality assurances associated with these products.
Specifications
Specifications
100 mL
B Cells, T Cells
Cell Isolation; In Vitro Diagnostic