Learn More
Rotaflo™ Straight Pattern Plain-side Arms for Burettes
Chemical resistant borosilicate glass with PTFE key and rotary design for fine single drop control
Brand:  Quickfit GP6ST/7
 View more versions of this product
Product Code. 10355240
  • 336.00 € / Pack of 5
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • No lubrication required - no seizure and no contamination
  • Temperature range: -20°C to 200°C
  • Vacuum capability to 10 Torr
  • Increased barrel diameter for drainage
  • Locking ring to prevent accidental removal
  • Ergonomic key for good grip and easy operation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plain Side-arm
10 mm
Borosilicate Glass
GP Rotaflo™ Stopcock