missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Rotarus™ PKF 60-24-3

Product Code. 16407049
Click to view available options
Capacity (Metric):
0.2 to 500 L/min
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16407049

Brand: Hirschmann™ 951106061

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

High quality PKF 60-series single-channel pump head that can be used in combination with stopper-less tubes.

  • 3 rollers
  • For WT 2.4 mm tubes
  • Only an adapter plate is necessary for the connection between pump head and pump drive
  • The pump head of the flow and volume pump drives is recognized by RFID technology
  • A simple and easy loading of the tubing is given by unlocking the pump head
  • The pump head is made of durable PPS, a high-performance plastic with very high heat resistance, high chemical resistance and durability

Specifications

Accuracy Approx. 5%
Capacity (Metric) 0.2 to 500 L/min
Color Black
Dimensions (L x W x H) 54 x 94 x 106 mm
Flow Rate Generally from 0.34 to 1400 mL/min. per channel
For Use With Pump drive and tubing
Material Polyphenylene Sulfide
No. of Rollers 3
Overall Height (Metric) 106 mm
Overall Length (Metric) 54 mm
Overall Width (Metric) 94 mm
Quantity 1
Roller Material Stainless Steel 304
Sterility Non-sterile
Case Material PPS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.