Learn More
Hirschmann Instruments™ rotarus™ Volume 100 Peristaltic Pump
Facilitates high flow rates conveniently and comprehensively in the laboratory. Hirschmann Instruments™ rotarus™ Volume 100 Peristaltic Pump offers continuous delivery in a dispensing pump across a broad range of applications.
Brand:  Hirschmann Instruments™ 9501562
Product Code. 15443583
  • 5680.00 € / Each
Explore more special offers
Description
Description
  • Changeable pump heads, single and multichannel design
  • Rapid tool-free changing of pump heads Automatic pump head and hose detection with RFID technology Control panel located directly on pump housing TFT touchscreen with user-friendly menu
  • Calibratable
  • Method memory
  • Operation with 24 V DC USB and RS232 interface for PC link using interface (available as accessory)
  • Pump heads from other manufacturers can be fitted without difficulty using an adapter
  • Comprehensive accessory range
Specifications
Specifications
1 to 12 (Depending on the pump head type used)
volume 100
270 x 100 x 220 mm
100/240 VAC
50/60 Hz
5 to 500 rpm
DC
10.1 kg
100 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products