Learn More
IKA™ Rotary Evaporator Lip Seal
For use with RV 10.8001 Rotary Evaporators.
Brand:  IKA™ 0003907000
Product Code. 11963817
  • 153.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • High chemical resistance
  • Manufactured from a PTFE compound
  • With a built-in stainless steel spring
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lip seal
Rotary Evaporators Lip-seal
Stainless Steel Spring
PTFE
RV 10.8001 rotary evaporators
1 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IKA™ Rotary Evaporator Lip Seal > For RV 10.8001