missing translation for 'onlineSavingsMsg'
Learn More

Welch ILMVAC™ Rotary Vane Pump

Withstands large amounts of water vapor in the vacuum pump’s inlet vapor stream. Welch ILMVAC™ Rotary Vane Pump is low maintenance, compact, and exceptionally quiet.

Brand:  Welch ILMVAC™ 322004

 View more versions of this product

Product Code. 10285789

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Two stage rotary vane pump
  • High water vapor tolerance (60 mbar for P6Z and 35 mbar for P12Z)
  • Max. leak rate of inlet valve 7x10-5 mbar l/sec
  • Integrated oil control glass
  • High temperature heat dissipation
  • Easy to service and clean
TRUSTED_SUSTAINABILITY
Specifications

Specifications

370 w
P6Z
96.7/110 L/min.
Rotary Vane Vacuum Pump
60 mbar (Water vapor tolerance)
150 x 415 x 210 mm
20 kg
50/60 Hz
Process Engineering
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates
View accessory products

Accessories and Services

Mandatory Accessory Optional Accessory Spare Part

Show More Show Less

Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Welch ILMVAC™ Rotary Vane Pump >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.