Learn More
Testo™ Rotating Display Compact Thermo Hygrometer
Product Code. 10344913
Click to view available options
Temperature Range (Metric):
0°C to 50°C
Unit Size:
Each
Product Code. 10344913
Brand: Testo™ 0560 6053
Only null left
Add to basket
Add to basket
Specifications
Type Compact Thermohygrometer
Temperature Range (Metric) 0°C to 50°C
Battery Type 3 AAA batteries
Display Type Rotating digital display
For Use With (Application) Measurement of air humidity and air temperature, as well as calculations of dewpoint temperature; Ideally suited for duct measurements
Includes testo 605-H1, 3 AAA batteries, attachment clip